Thermo Scientific Chemicals

Ammonium formate, 98+%, Thermo Scientific Chemicals

Catalog number: 014517.18
50 g, Each
Thermo Scientific Chemicals

Ammonium formate, 98+%, Thermo Scientific Chemicals

Catalog number: 014517.18
50 g, Each
Quantity
Catalog number: 014517.18
also known as 014517-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-69-2
IUPAC Name
formic acid amine
Molecular Formula
CH5NO2
InChI Key
VZTDIZULWFCMLS-UHFFFAOYSA-N
SMILES
N.OC=O
Form
Deliquescent powder/granule
Assay (Titration)
>98.0%
Chloride content
≤5 ppm
Heavy Metals
≤0.001%
Residue on ignition
≤0.005%

Description

Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text