Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, ACS, 99.5-101.0%, Thermo Scientific Chemicals

Catalog number: 014518.A3
2 kg, Each
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, ACS, 99.5-101.0%, Thermo Scientific Chemicals

Catalog number: 014518.A3
2 kg, Each
Quantity
Catalog number: 014518.A3
also known as 014518-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-17-7
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Formula
H10Na2O8S2
InChI Key
PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
Assay (unspecified)
99.5-101.0% Na₂O₃S₂.5H₂O
Impurity content
Sulfate and sulfite (as SO₄) 0.1%
Impurity content
Sulfide P.T. (limit about 1ppm)
Impurity content
Nitrogen compounds (as N) 0.002%
Insoluble material
0.005%

Description

Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivatives, S-alkylthiosulfonates, are known as Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solutions and to check the pH of bleach solutions with liquid indicators. Other uses include tanning of leather; as an active component in hand warmers and chemical heating pads and as an anionic surfactant to aid in the dispersion of pharmaceutical preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text