Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 014535.06
also known as 014535-06
Price (USD)/ Each
741.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
741.00
Add to cart
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Catalog number014535.06
Price (USD)/ Each
741.00
-
Add to cart
Specifications
Chemical Name or MaterialBis(cyclopentadienyl)manganese
Name NoteSublimed
CAS73138-26-8
Health Hazard 1H228-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only