Thermo Scientific Chemicals

Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 014537.06
5 g, Each
Thermo Scientific Chemicals

Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 014537.06
5 g, Each
Quantity
Catalog number: 014537.06
also known as 014537-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-35-8
IUPAC Name
lithium(1+) bromide
Molecular Formula
BrLi
InChI Key
AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES
[Li+].[Br-]
Residual water
< 100 ppm
Total Metal Impurities
0.002% max.
Particle Size
-10 Mesh
Appearance (Color)
White
Form
Beads

Description

Lithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text