Thermo Scientific Chemicals

Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014562.18
50 g, Each
Thermo Scientific Chemicals

Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014562.18
50 g, Each
Quantity
Catalog number: 014562.18
also known as 014562-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13266-82-5
IUPAC Name
diyttrium(3+) nonahydrate trioxalate
Molecular Formula
C6H18O21Y2
InChI Key
CTXJKXXQOORSKU-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.O.O.O.[Y+3].[Y+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Form
Powder
Assay from Suppliers CofA
Report rare earth impurities
Appearance (Color)
White
Assay from Suppliers CofA
≥99.9% REO

Description

Yttrium oxalate is applied in ceramics, glass, and electronics. It is an important material for tri-bands rare Earth phosphors and yttrium-iron-garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text