Thermo Scientific Chemicals

Lutetium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014568.14
25 g, Each
Thermo Scientific Chemicals

Lutetium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014568.14
25 g, Each
Quantity
Catalog number: 014568.14
also known as 014568-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100641-16-5
IUPAC Name
lutetium(3+) trinitrate
Molecular Formula
LuN3O9
InChI Key
APRNQTOXCXOSHO-UHFFFAOYSA-N
SMILES
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lutetium(III) nitrate hydrate is used in ceramics, glass, phosphors and in lasers. It finds application as a catalyst in petroleum cracking and to prepare laser crystal. Further, it is used in hydrogenation, alkylation and polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text