Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 014574.18
also known as 014574-18
Price (USD)/ Each
343.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
343.00
Add to cart
Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number014574.18
Price (USD)/ Each
343.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialYtterbium(III) carbonate hydrate
Melting Point(decomposition)
Recommended StorageAmbient temperatures
Physical FormCrystalline
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only