Thermo Scientific Chemicals

Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014576.09
10 g, Each
Thermo Scientific Chemicals

Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014576.09
10 g, Each
Quantity
Catalog number: 014576.09
also known as 014576-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15280-58-7
IUPAC Name
ytterbium(3+) triacetate
Molecular Formula
C6H9O6Yb
InChI Key
OSCVBYCJUSOYPN-UHFFFAOYSA-K
SMILES
[Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Yttrium Acetate is widely applied in electronic ceramics, glass, and electronics. Yttrium compounds are used as a catalyst for ethylene polymerization. Yttrium is used in the production of a large variety of synthetic garnets, and Yttria is used to make Yttrium Iron Garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text