Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 014576.18
also known as 014576-18
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
Add to cart
Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number014576.18
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15280-58-7
IUPAC Nameytterbium(3+) triacetate
Molecular FormulaC6H9O6Yb
InChI KeyOSCVBYCJUSOYPN-UHFFFAOYSA-K
SMILES[Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Yttrium Acetate is widely applied in electronic ceramics, glass, and electronics. Yttrium compounds are used as a catalyst for ethylene polymerization. Yttrium is used in the production of a large variety of synthetic garnets, and Yttria is used to make Yttrium Iron Garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only