Thulium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thulium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thulium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 014579.18
also known as 014579-18
Price (USD)/ Each
1,037.65
Online exclusive
1153.00 
Save 115.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
1,037.65
Online exclusive
1153.00 
Save 115.35 (10%)
Add to cart
Thulium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number014579.18
Price (USD)/ Each
1,037.65
Online exclusive
1153.00 
Save 115.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialThulium(III) nitrate hydrate
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
Recommended StorageAmbient temperatures
View more
Thulium(III) nitrate hydrate is used as a laboratory reagent. It finds application in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only