Thermo Scientific Chemicals

Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014588.09
10 g, Each
Thermo Scientific Chemicals

Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014588.09
10 g, Each
Quantity
Catalog number: 014588.09
also known as 014588-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14483-18-2
IUPAC Name
holmium(3+) pentahydrate trinitrate
Molecular Formula
H10HoN3O14
InChI Key
BJUGWWDCFYEYOA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Ho+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text