Thermo Scientific Chemicals

Terbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014600.18
50 g, Each
Thermo Scientific Chemicals

Terbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014600.18
50 g, Each
Quantity
Catalog number: 014600.18
also known as 014600-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100587-92-6
IUPAC Name
terbium(3+) triacetate
Molecular Formula
C6H9O6Tb
InChI Key
JQBILSNVGUAPMM-UHFFFAOYSA-K
SMILES
[Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Terbium Acetate has important role as an activator for green phosphors used in color TV tubes. Meanwhile Terbium Acetate is also used in special lasers and as a dopant in solid-state devices. Terbium is also used in alloys and in the production of electronic devices. As a component of Terfenol-D, Terbium is of use in actuators, in naval sonar systems, sensors, in the SoundBugdevice (its first commercial application), and other magnetomechanical devices. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text