Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014601.14
also known as 014601-14
Price (USD)/ Each
45.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
45.60
Add to cart
Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number014601.14
Price (USD)/ Each
45.60
-
Add to cart
Chemical Identifiers
CAS13465-58-2
IUPAC Namedisamarium(3+) trisulfate
Molecular FormulaO12S3Sm2
InChI KeyLVSITDBROURTQX-UHFFFAOYSA-H
SMILES[Sm+3].[Sm+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only