Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 014603.22
also known as 014603-22
Price (USD)/ Each
140.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
140.00
Add to cart
Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number014603.22
Price (USD)/ Each
140.00
-
Add to cart
Chemical Identifiers
CAS13517-12-9
IUPAC Namesamarium(3+) hexahydrate tribromide
Molecular FormulaBr3H12O6Sm
InChI KeyLXJDCZVUWJNXRL-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only