Thermo Scientific Chemicals

Samarium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014604.22
100 g, Each
Thermo Scientific Chemicals

Samarium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014604.22
100 g, Each
Quantity
Catalog number: 014604.22
also known as 014604-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17829-86-6
IUPAC Name
samarium(3+) triacetate
Molecular Formula
C6H9O6Sm
InChI Key
JPDBEEUPLFWHAJ-UHFFFAOYSA-K
SMILES
[Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Samarium Acetate has specialized uses in glass, phosphors, lasers, and thermoelectric devices. Another important application of Samarium and its compounds is as catalyst and chemical reagent. One of the most important applications of Samarium is in Samarium-Cobalt magnets. . These magnets are found in small motors, headphones, and high-end magnetic pickups for guitars and related musical instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text