Thermo Scientific Chemicals

Indium antimonide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014626.04
2 g, Each
Thermo Scientific Chemicals

Indium antimonide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014626.04
2 g, Each
Quantity
Catalog number: 014626.04
also known as 014626-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-41-0
Total Metal Impurities
0.001% max.

Description

Indium antimonide finds use in infrared detectors, including FLIR systems, thermal imaging cameras, infrared astronomy and in infrared homing missile guidance systems, in fast transistors. It is used in thermal image detectors using photo-electromagnetic detectors or photodiodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text