Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis), Thermo Scientific Chemicals
Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 014629.A1
also known as 014629-A1
Price (USD)/ Each
83.30
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
83.30
Add to cart
Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis), Thermo Scientific Chemicals
Catalog number014629.A1
Price (USD)/ Each
83.30
-
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Appearance (Color)Gray
Comment2.5 cm and down
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only