Titanium(IV) oxide, rutile, 99.99% (metals basis)
Titanium(IV) oxide, rutile, 99.99% (metals basis)
Titanium(IV) oxide, rutile, 99.99% (metals basis)
Titanium(IV) oxide, rutile, 99.99% (metals basis)
Titanium(IV) oxide, rutile, 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium(IV) oxide, rutile, 99.99% (metals basis)

CAS: 1317-80-2 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 014631.30
also known as 014631-30
Price (USD)
1,505.00
Each
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,505.00
Each
Add to cart
Chemical Identifiers
CAS1317-80-2
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Ask our AI about this Product
Titanium(IV) oxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link