Silver flake, 80% <20 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Silver flake, 80% &lt;20 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver flake, 80% <20 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 014639.18
also known as 014639-18
Price (USD)/ Each
244.65
Online exclusive
272.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
244.65
Online exclusive
272.00 
Save 27.35 (10%)
Add to cart
Silver flake, 80% <20 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014639.18
Price (USD)/ Each
244.65
Online exclusive
272.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
CommentScott density 18-28g/in³
Surface Area1.0-2.2m²/g
Total Metal Impurities0.1% max.
FormFlake
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only