Thermo Scientific Chemicals

Tantalum(V) ethoxide, 99.999% (metals basis), Nb <100ppm, Thermo Scientific Chemicals

Catalog number: 014643.03
1 g, Each
Thermo Scientific Chemicals

Tantalum(V) ethoxide, 99.999% (metals basis), Nb <100ppm, Thermo Scientific Chemicals

Catalog number: 014643.03
1 g, Each
Quantity
Catalog number: 014643.03
also known as 014643-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6074-84-6
IUPAC Name
pentaethoxytantalum
Molecular Formula
C10H25O5Ta
InChI Key
HSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILES
CCO[Ta](OCC)(OCC)(OCC)OCC
Appearance (Color)
Clear colorless to pale yellow
Assay from Supplier's CofA
≥99.5% (gravimetric)
Trace Metals
Nb: ≤100ppm
Form
Low melting solid or liquid
Total Metal Impurities
≤10ppm (0.001%) excluding Nb

Description

Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text