Thermo Scientific Chemicals

Copper(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014644.22
100 g, Each
Thermo Scientific Chemicals

Copper(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014644.22
100 g, Each
Quantity
Catalog number: 014644.22
also known as 014644-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-89-6
IUPAC Name
λ¹-copper(1+) chloride
Molecular Formula
ClCu
InChI Key
OXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES
[Cl-].[Cu+]
Total Metal Impurities
0.001% max.

Description

Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text