Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014652.AP
also known as 014652-AP
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
481.00 
Save 72.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
481.00 
Save 72.35 (15%)
Add to cart
Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Catalog number014652.AP
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
481.00 
Save 72.35 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPrecious Metals, plasma standard solution
ConcentrationMatrix: 20% HCl
Name NoteAu, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Quantity500 mL
View more
Precious Metals, plasma standard solution is useful as a standard for the determination of metals by inductively coupled plasma-mass spectrometry.