Thermo Scientific Chemicals
Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL, Thermo Scientific Chemicals
Catalog number: 014657.AE
100 mL, Each
Thermo Scientific Chemicals
Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL, Thermo Scientific Chemicals
Catalog number: 014657.AE
100 mL, Each
Quantity
Catalog number: 014657.AE
also known as 014657-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
Ag 100?g/mL ?2%
Concentration
Pb 100?g/mL ?2%
Concentration
Se 100?g/mL ?2%
Concentration
Ba 100?g/mL ?2%
Concentration
Cd 100?g/mL ?2%
Description
Heavy Metals, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.