Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
25 cm
5 m
Catalog number 014658.G1
also known as 014658-G1
Price (USD)/ Each
131.00
-
Add to cart
Quantity:
1 m
Request bulk or custom format
Price (USD)/ Each
131.00
Add to cart
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Catalog number014658.G1
Price (USD)/ Each
131.00
-
Add to cart
Specifications
Chemical Name or MaterialMolybdenum wire, platinum coated
Name Note0.457mm (0.018 in.) dia.
CAS7439-98-7
Recommended StorageAmbient temperatures
EINECS Number231-107-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only