Barium chromate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium chromate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 014669.30
also known as 014669-30
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
80.70 
Save 12.05 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
80.70 
Save 12.05 (15%)
Add to cart
Barium chromate, 98%, Thermo Scientific Chemicals
Catalog number014669.30
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
80.70 
Save 12.05 (15%)
-
Add to cart
Chemical Identifiers
CAS10294-40-3
IUPAC Namebarium(2+) dioxochromiumbis(olate)
Molecular FormulaBaCrO4
InChI KeyQFFVPLLCYGOFPU-UHFFFAOYSA-N
SMILES[Ba++].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder or crystals
Assay from Suppliers CofA≥97.5%
Appearance (Color)Pale yellow to yellow
Barium chromate is used as a pigment, oxidizing agent, colorant in glass, and sulfate scavenger in chromium electroplating baths, ceramics, and porcelain. It acts as a corrosion inhibitor in jointing pastes and metal primers. Further, it is used in fuses, safety matches, ignition control devices and high-temperature batteries. In addition to this, it is used as a carrier for the chromium ions and useful as a burn rate modifier in pyrotechnic compositions. It is involved as a catalyst in the alkane dehydration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only