Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

For Vacuum Deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 014684.14
also known as 014684-14
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number014684.14
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only