Thermo Scientific Chemicals

Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals

Catalog number: 014685.A1
1 kg, Each
Thermo Scientific Chemicals

Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals

Catalog number: 014685.A1
1 kg, Each
Quantity
Catalog number: 014685.A1
also known as 014685-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6556-16-7
IUPAC Name
manganese(2+) dihydrate oxalate
Molecular Formula
C2H4MnO6
InChI Key
HDJUVFZHZGPHCQ-UHFFFAOYSA-L
SMILES
O.O.[Mn++].[O-]C(=O)C([O-])=O
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
Manganese, Mn: ≥30%
Appearance (Color)
White to pale pink

Description

Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text