Thermo Scientific Chemicals

Nickel(II) chloride, anhydrous, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014687.22
100 g, Each
Thermo Scientific Chemicals

Nickel(II) chloride, anhydrous, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014687.22
100 g, Each
Quantity
Catalog number: 014687.22
also known as 014687-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7718-54-9
IUPAC Name
nickel(2+) dichloride
Molecular Formula
Cl2Ni
InChI Key
QMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ni++]
Total Metal Impurities
1% max.

Description

Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text