Thermo Scientific Chemicals

Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals

Catalog number: 014689.04
2 g, Each
Thermo Scientific Chemicals

Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals

Catalog number: 014689.04
2 g, Each
Quantity
Catalog number: 014689.04
also known as 014689-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3236-82-6
IUPAC Name
pentaethoxyniobium
Molecular Formula
C10H25NbO5
InChI Key
ZTILUDNICMILKJ-UHFFFAOYSA-N
SMILES
CCO[Nb](OCC)(OCC)(OCC)OCC
Form
Liquid
Elemental Analysis
Ta: ≤500ppm
Total Metal Impurities
≤10ppm ( excluding Ta)
Appearance (Color)
Orange

Description

Preparation of niobium oxide and mixed metal oxides.Niobium(V) ethoxide is used for the sol-gel processing of materials containing niobium oxides and mixed metal oxides. It is also used to prepare ferroelectric nanomaterials consisting of oxides of potassium, silicon and niobium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text