Thermo Scientific Chemicals

Manganese(II) chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 014697.14
25 g, Each
Thermo Scientific Chemicals

Manganese(II) chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 014697.14
25 g, Each
Quantity
Catalog number: 014697.14
also known as 014697-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7773-01-5
IUPAC Name
manganese(2+) dichloride
Molecular Formula
Cl2Mn
InChI Key
GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Mn++]
Total Metal Impurities
0.002% max.
Appearance (Color)
Pink
Form
Beads
Residual water
< 100 ppm
Particle Size
-10 Mesh

Description

Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It is also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text