Thermo Scientific Chemicals

Potassium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014700.22
100 g, Each
Thermo Scientific Chemicals

Potassium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014700.22
100 g, Each
Quantity
Catalog number: 014700.22
also known as 014700-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Total Metal Impurities
0.1% max.
Form
Beads
Residual water
< 100 ppm
Appearance (Color)
White
Particle Size
-10 Mesh

Description

Potassium bromide is widely used as an anticonvulsant and a sedative. Also used in the preparation of stable Pd nanocubes that si used in immunolabeling. Potassium bromide is used for the purification of Sarcocystis neurona sporocysts by discontinuous density gradient centrifugation. May be used for removal of peripheral membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text