Thermo Scientific Chemicals

Silicon(IV) nitride, alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014704.22
100 g, Each
Thermo Scientific Chemicals

Silicon(IV) nitride, alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014704.22
100 g, Each
Quantity
Catalog number: 014704.22
also known as 014704-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12033-89-5
IUPAC Name
Silicon nitride
Molecular Formula
N4Si3
InChI Key
HQVNEWCFYHHQES-UHFFFAOYSA-N
SMILES
N12[Si]34N5[Si]11N3[Si]25N41
Total Metal Impurities
0.1% max.

Description

Silicon(IV) nitride is used in automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from Silicon(IV) nitride show good shock resistance. Similarly, bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and is used in the manufacturing of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text