Thermo Scientific Chemicals

Sodium hydrogen carbonate, ACS, 99.7-100.3%, Thermo Scientific Chemicals

Catalog number: 014707.A3
2 kg, Each
Thermo Scientific Chemicals

Sodium hydrogen carbonate, ACS, 99.7-100.3%, Thermo Scientific Chemicals

Catalog number: 014707.A3
2 kg, Each
Quantity
Catalog number: 014707.A3
also known as 014707-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-55-8
IUPAC Name
sodium hydrogen carbonate
Molecular Formula
CHNaO3
InChI Key
UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES
[Na+].OC([O-])=O
Assay (unspecified)
99.7-100.3% NaHCO₃ min
Impurity content
Fe 0.001%
Impurity content
K 0.005%
Insoluble material
0.015%
Heavy Metals
(ICP-OES) 5ppm

Description

Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and as a pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, in baking powder, as a mild abrasive and a cleaning compound. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text