Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals
Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 014709.09
also known as 014709-09
Price (USD)/ Each
44.30
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
44.30
Add to cart
Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals
Catalog number014709.09
Price (USD)/ Each
44.30
-
Add to cart
Chemical Identifiers
CAS1314-61-0
IUPAC Name[(dioxotantalio)oxy]dioxotantalum
Molecular FormulaO5Ta2
InChI KeyPBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILESO=[Ta](=O)O[Ta](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Total Metal Impurities≤1%
Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only