Thermo Scientific Chemicals

Titanium(IV) chloride, 99.0% min, typically 99.6%, Thermo Scientific Chemicals

Catalog number: 014713.K2
1 L, Each
Thermo Scientific Chemicals

Titanium(IV) chloride, 99.0% min, typically 99.6%, Thermo Scientific Chemicals

Catalog number: 014713.K2
1 L, Each
Quantity
Catalog number: 014713.K2
also known as 014713-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-45-0
IUPAC Name
titanium(4+) tetrachloride
Molecular Formula
Cl4Ti
InChI Key
XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (unspecified)
≥99.0%
Assay (unspecified)
Report metals analysis

Description

In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text