Thermo Scientific Chemicals

Copper(I) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014718.A1
1 kg, Each
Thermo Scientific Chemicals

Copper(I) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014718.A1
1 kg, Each
Quantity
Catalog number: 014718.A1
also known as 014718-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22205-45-4
IUPAC Name
bis(λ¹-copper(1+)) sulfanediide
Molecular Formula
Cu2S
InChI Key
AQMRBJNRFUQADD-UHFFFAOYSA-N
SMILES
[S--].[Cu+].[Cu+]
Total Metal Impurities
0.5% max.

Description

Copper(I) sulfide is used in luminous paints, electrodes and in solar cells. It is also used for grain mounts, XRD and in microprobe standards for the identification of unknown minerals. It acts as a precursor for the preparation of copper(I) oxide and copper metal. Further, it acts as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text