Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 014720.18
also known as 014720-18
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
Add to cart
Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number014720.18
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only