Gold wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Gold wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Form: Approx.3.79g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
10 cm
250 cm
Catalog number 014728.BW
also known as 014728-BW
Price (USD)/ Each
470.65
Online exclusive
523.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
50 cm
Request bulk or custom format
Price (USD)/ Each
470.65
Online exclusive
523.00 
Save 52.35 (10%)
Add to cart
Gold wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number014728.BW
Price (USD)/ Each
470.65
Online exclusive
523.00 
Save 52.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Diameter0.45-0.55mm
Total Metal Impurities≤ 0.05%
FormWire
CommentGold
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only