Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 cm
5 cm
25 cm
Catalog number 014729.BO
also known as 014729-BO
Price (USD)/ Each
182.00
-
Add to cart
Quantity:
1 cm
Request bulk or custom format
Price (USD)/ Each
182.00
Add to cart
Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number014729.BO
Price (USD)/ Each
182.00
-
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.05%
FormWire, 2.0 mm (0.08 in.) dia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only