Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
113 g
28 g
227 g
454 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014735.TB
Price (USD)/ Each
-
Quantity:
113 g
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGraphite powder
Name NoteNatural
CAS7782-42-5
Health Hazard 1H228-H319-H335
Health Hazard 2GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only