Thermo Scientific Chemicals
Graphite powder, natural, microcrystal grade, APS 2-15 micron, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number: 014736.TB
113 g, Each
Thermo Scientific Chemicals
Graphite powder, natural, microcrystal grade, APS 2-15 micron, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number: 014736.TB
113 g, Each
Quantity
Catalog number: 014736.TB
Price (USD)
Have Questions?
Total Metal Impurities
0.0005% max.
Description
Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.