Thermo Scientific Chemicals

Graphite electrode, crater-drillpoint/undercut, 6.15mm dia, 38.10mm length, volume 0.072cc, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 014743.KN
100 Pc., Tube, Each
Thermo Scientific Chemicals

Graphite electrode, crater-drillpoint/undercut, 6.15mm dia, 38.10mm length, volume 0.072cc, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 014743.KN
100 Pc., Tube, Each
Quantity
Packaging
Catalog number: 014743.KN
Price (USD)
Have Questions?
Assay from Supplier's CofA
≥99.9995% (metals basis)
Length
38.10mm length
Volume
0.072cc volume
Appearance (Color)
Grey
Form
Electrode, crater-drillpoint/undercut, 6.15mm diameter.

Description

Graphite electrodes are used mainly in electric arc furnace steel production. It also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings, charging practice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text