Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Thermo Scientific Chemicals
Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
7.88 cc
7.5 cc
Shape:
Drillpoint
Quantity:
View all
36pc
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014747.UB
Price (USD)/ Each
-
Capacity (Metric):
7.88 cc
Shape:
Drillpoint
Quantity:
36pc
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
DescriptionUnpurified extruded graphite
MaterialGraphite
For Use With (Application)Metallurgy
Quantity36pc
Capacity (Metric)7.88 cc