Thermo Scientific Chemicals

Graphite electrode, counter-flat top, 3.05mm dia, 38.10 mm long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 014754.KN
100 Pc., Tube, Each
Thermo Scientific Chemicals

Graphite electrode, counter-flat top, 3.05mm dia, 38.10 mm long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 014754.KN
100 Pc., Tube, Each
Quantity
Packaging
Catalog number: 014754.KN
Price (USD)
Have Questions?
Appearance (Color)
Grey
Form
Electrode, counter-flat top, 3.05mm diameter.
Assay from Supplier's CofA
≥99.9995% (metals basis)
Length
38.10mm length

Description

It is used as homogeneous catalysts. They are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Graphite is used widely as an engineering material over a variety of applications. Applications include piston rings, thrust bearings, journal bearings and vanes. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text