Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals
Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014756.FF
Price (USD)/ Each
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-41-7
IUPAC Nameberyllium
Molecular FormulaBe
InChI KeyATBAMAFKBVZNFJ-UHFFFAOYSA-N
SMILES[Be]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99% (metals basis)
FormFoil, 0.5mm (0.02in) thick.
Appearance (Color)Dark gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only