Thermo Scientific Chemicals

Deuterium oxide, 99.8% (Isotopic), Thermo Scientific Chemicals

Catalog number: 014764.30
250 g, Each
Thermo Scientific Chemicals

Deuterium oxide, 99.8% (Isotopic), Thermo Scientific Chemicals

Catalog number: 014764.30
250 g, Each
Quantity
Catalog number: 014764.30
also known as 014764-30
Price (EUR)

Chemical Identifiers

CAS
7789-20-0
IUPAC Name
[(²H)oxy](²H)
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES
[2H]O[2H]
Appearance (Color)
Clear colorless
Form
Liquid
Proton NMR
Chemical Purity: ≥99.5% (Passes Test)
Proton NMR
Isotropic Enrichment: ≥99.8%

Description

Deuterium oxide is used in nuclear magnetic resonance spectroscopy (NMR). It is also useful in the identification of labile hydrogens. As a source of deuterium, it is utilized for preparing specifically labeled isotopologs of organic compounds. It is often used as a substitute for water in the analysis of proteins in solution by using fourier transform infrared spectroscopy (FTIR). It finds application in certain types of nuclear reactors and in tritium production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text