Ethanesulfonic acid, 96%, Thermo Scientific Chemicals
Ethanesulfonic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanesulfonic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 014891.30
also known as 014891-30
Price (USD)/ Each
596.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
596.00
Add to cart
Ethanesulfonic acid, 96%, Thermo Scientific Chemicals
Catalog number014891.30
Price (USD)/ Each
596.00
-
Add to cart
Chemical Identifiers
CAS594-45-6
IUPAC Nameethanesulfonate
Molecular FormulaC2H5O3S
InChI KeyCCIVGXIOQKPBKL-UHFFFAOYSA-M
SMILESCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Aqueous acid-base Titration)≥95.0%
Ethanesulfonic acid is employed in the electrolytic reduction of perrhenate solutions. It is also used as a catalyst for alkylation, polymerization and other reactions. Further, it reacts with triethoxymethane to prepare ethanesulfonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only