Thermo Scientific Chemicals

1-Chloroadamantane, 98%, Thermo Scientific Chemicals

Catalog number: 015169.22
100 g, Each
Thermo Scientific Chemicals

1-Chloroadamantane, 98%, Thermo Scientific Chemicals

Catalog number: 015169.22
100 g, Each
Quantity
Catalog number: 015169.22
also known as 015169-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
935-56-8
IUPAC Name
1-chloroadamantane
Molecular Formula
C10H15Cl
InChI Key
OZNXTQSXSHODFR-UHFFFAOYSA-N
SMILES
ClC12CC3CC(CC(C3)C1)C2
Assay (Titration)
≥97.5%
Form
Crystalline powder / chunks
Appearance (Color)
White to pale grey

Description

It is used in some dry etching masks and polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text