Thermo Scientific Chemicals

Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 015290.04
2 g, Each
Thermo Scientific Chemicals

Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 015290.04
2 g, Each
Quantity
Catalog number: 015290.04
also known as 015290-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-53-5
IUPAC Name
europium(3+) hexahydrate trinitrate
Molecular Formula
EuH12N3O15
InChI Key
JVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Europium(III) nitrate hexahydrate is used in the preparation of pharmaceuticals, agrochemicals and other chemical products. It is used as a catalyst and a component of specialized steels. It is also used in the synthesis of light emitting nanomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text