Thermo Scientific Chemicals

Cerium(III) acetate sesquihydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 015294.36
500 g, Each
Thermo Scientific Chemicals

Cerium(III) acetate sesquihydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 015294.36
500 g, Each
Quantity
Catalog number: 015294.36
also known as 015294-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17829-82-2
Total Rare Earth Oxide Impurities
0.1% max.

Description

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text